Tlačiť
ESD safety ankle boots, lace-up, Impulse EA blue Mid, S2
Dodávateľ: ELTEN
7672603/41 7672603/42 7672601/40 7672601/41 7672601/42 7672601/43 7672601/44 7672601/45 7672601/46 7672601/47 7672601/48 7672602/40 7672602/41 7672602/42 7672602/43 7672602/44 7672602/45 7672602/46 7672602/47 7672602/48 7672603/40 7672603/43 7672603/44 7672603/45 7672603/46 7672603/47 7672603/48
ELTN7672603/41EA 113.12 EUR
ELTN7672603/41 ELTN7672603/42 ELTN7672601/40 ELTN7672601/41 ELTN7672601/42 ELTN7672601/43 ELTN7672601/44 ELTN7672601/45 ELTN7672601/46 ELTN7672601/47 ELTN7672601/48 ELTN7672602/40 ELTN7672602/41 ELTN7672602/42 ELTN7672602/43 ELTN7672602/44 ELTN7672602/45 ELTN7672602/46 ELTN7672602/47 ELTN7672602/48 ELTN7672603/40 ELTN7672603/43 ELTN7672603/44 ELTN7672603/45 ELTN7672603/46 ELTN7672603/47 ELTN7672603/48
ESD safety ankle boots, lace-up, Impulse EA blue Mid, S2
Obuv
Leather-free, sporty and lightweight ESD safety ankle boots made from microfibre, reflective materials, a full padded bellows tongue and a slip resistant dual-density TPU/PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Full-length semi-orthopaedic inlay sole ESD black
  • Soft fleece ESD insole
  • Protective steel toecap (200 J)
  • Abrasion proof toe protection
Ideal for use in areas where exposure to moisture is expected
Colour: Blue/black
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifikácie: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC, form B