Tlačiť
ESD safety ankle boots, lace-up, Matthew Pro GTX Mid
Dodávateľ: ELTEN
7678101/40 7678101/41 7678101/42 7678101/43 7678101/44 7678101/45 7678101/46 7678101/47 7678101/48 7678102/40 7678102/41 7678102/42 7678102/43 7678102/44 7678102/45 7678102/46 7678102/47 7678102/48 7678103/40 7678103/41 7678103/42 7678103/43 7678103/44 7678103/45 7678103/46 7678103/47 7678103/48 1767801/36 1767801/37 1767801/38 1767801/39 1767802/36 1767802/37 1767802/38 1767802/39 1767803/36 1767803/37 1767803/38 1767803/39
ELTN7678101/40EA 137.59 EUR
ELTN7678101/40 ELTN7678101/41 ELTN7678101/42 ELTN7678101/43 ELTN7678101/44 ELTN7678101/45 ELTN7678101/46 ELTN7678101/47 ELTN7678101/48 ELTN7678102/40 ELTN7678102/41 ELTN7678102/42 ELTN7678102/43 ELTN7678102/44 ELTN7678102/45 ELTN7678102/46 ELTN7678102/47 ELTN7678102/48 ELTN7678103/40 ELTN7678103/41 ELTN7678103/42 ELTN7678103/43 ELTN7678103/44 ELTN7678103/45 ELTN7678103/46 ELTN7678103/47 ELTN7678103/48 ELTN1767801/36 ELTN1767801/37 ELTN1767801/38 ELTN1767801/39 ELTN1767802/36 ELTN1767802/37 ELTN1767802/38 ELTN1767802/39 ELTN1767803/36 ELTN1767803/37 ELTN1767803/38 ELTN1767803/39
ESD safety ankle boots, lace-up, Matthew Pro GTX Mid
Obuv
Leather-free ESD safety ankle boots are made from hydrophobised microfibre and hydrophobised textile material CORDURA® leather with GORE-TEX performance, reflective materials, a fully padded bellows tongue and a dual densitiy PU/PU sole ERGO-ACTIVE 2.
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Semi-orthopaedic inlay sole ESD black
  • Anatomically-shaped insole and ankle support for improved stability
  • Abrasion resistant scuff cap protects the upper
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects. The sizes 36 to 39 are ideally tailored to the ergonomics of female feet.
Colour: Black/red
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifikácie: DGUV 112-191; EN 61340-5-1 (ESD); EN ISO 20345; S3, CI, SRC, Form B