Tlačiť
ESD safety ankle boots, lace-up, DANIEL Pro Mid
Dodávateľ: ELTEN
7679101/40 7679101/41 7679101/42 7679101/43 7679101/44 7679101/45 7679101/46 7679101/47 7679101/48 7679102/40 7679102/41 7679102/42 7679102/43 7679102/44 7679102/45 7679102/46 7679102/47 7679102/48 7679103/40 7679103/41 7679103/42 7679103/43 7679103/44 7679103/45 7679103/46 7679103/47 7679103/48
ELTN7679101/40EA 116.73 EUR
ELTN7679101/40 ELTN7679101/41 ELTN7679101/42 ELTN7679101/43 ELTN7679101/44 ELTN7679101/45 ELTN7679101/46 ELTN7679101/47 ELTN7679101/48 ELTN7679102/40 ELTN7679102/41 ELTN7679102/42 ELTN7679102/43 ELTN7679102/44 ELTN7679102/45 ELTN7679102/46 ELTN7679102/47 ELTN7679102/48 ELTN7679103/40 ELTN7679103/41 ELTN7679103/42 ELTN7679103/43 ELTN7679103/44 ELTN7679103/45 ELTN7679103/46 ELTN7679103/47 ELTN7679103/48
ESD safety ankle boots, lace-up, DANIEL Pro Mid
Obuv
ESD safety ankle boots made from hydrophobised nubuck leather material, fabric lining, reflective materials, a full padded bellows tongues, TPU scuff cap and a dual density PU/PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Semi-orthopaedic inlay sole ESD black
  • Metal-free, puncture resistant midsole
  • Anatomically-shaped insole and ankle support for improved stability
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifikácie: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, form B