Tlačiť
ESD safety sandals, Velcro® fastening, Wellmaxx, Impulse XXT Easy
Dodávateľ: ELTEN
719755/36 719755/37 719755/38 719755/39 719755/40 719755/41 719755/42 719755/43 719755/44 719755/45 719755/46 719755/47 719755/48
ELTN719755/36EA 109.51 EUR
ELTN719755/36 ELTN719755/37 ELTN719755/38 ELTN719755/39 ELTN719755/40 ELTN719755/41 ELTN719755/42 ELTN719755/43 ELTN719755/44 ELTN719755/45 ELTN719755/46 ELTN719755/47 ELTN719755/48
ESD safety sandals, Velcro® fastening, Wellmaxx, Impulse XXT Easy
Obuv
Metal and leather free, super lightweight ESD safety shoes made from microfibre upper, reflective materials, a padded tongue and a dual density PU/PU sole WELLMAXX TRAINERS.
  • Fabric lining for natural ventilation
  • Full-length inlay sole ESD PRO black
  • Soft fleece ESD insole
  • Anatomically shaped insole and ankle support for improved stability
  • Cushioning midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving the energy back to the wearer
  • Protective composite toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/blue
Certifikácie: EN 61340-5-1 (ESD), EN ISO 20345; S1, SRC, form A