Tlačiť
Knitted gloves, uvex unipur carbon
Dodávateľ: UVEX
6055606 6055607 6055608 6055609 6055610
112-5417EA 7.49 EUR
112-5417 112-5418 112-5419 112-5420 112-5421
Knitted gloves, uvex unipur carbon
Rukavice
These polyamide gloves with knitted cuff, thin elastomer coating on the fingertips and carbon micro-nubbing on the palm are sensitive and antistatic.
  • Excellent grip
  • Very high breathability
  • Outstanding tactile feel
Applications: Working with touchscreens, electrical industry, work in anti-static areas, assembly of electronic components.
Certifikácie: DIN EN 16350:2014; EN 388:2016 (2131X)