Tlačiť

Ricinoleic acid ≥99% (by GC), Sigma-Aldrich®

Dodávateľ: Merck

Synonymá: Ricinolic acid, (R)-12-Hydroxy-cis-9-octadecenoic acid, 12-Hydroxyoleic acid, Ricinelaidic acid, (R,Z)-12-Hydroxyoctadec-9-enoic acid

M-Clarity™ quality level = MQ200

R7257-100MG R7257-1G
SIALR7257-100MGEA 181.45 EUR
SIALR7257-100MG SIALR7257-1G
Ricinoleic acid ≥99% (by GC), Sigma-Aldrich®
Ricinoleic acid
Vzorec: C₁₈H₃₄O₃
Mw: 298.47 g/mol
Hustota: 0.940 g/ml (20 °C) (lit.)
Teplota vzplanutia: 224 °C
Skladovacia teplota: Freezer
Číslo MDL: MFCD00084840
Číslo CAS: 141-22-0

Order Now

Výsledky špecifikačného testu

Appearance (Color) Colorless to Very Faint Yellow
Appearance (Turbidity) Clear
Appearance (Form) At 37 °C to 40 °C Liquid
Proton NMR Spectrum Conforms to Structure
Purity (GC) ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR