Antibody labelling kits, Mix-n-Stain™ DNP
Dodávateľ: Biotium
92326
92327
92325
92326.EA
255.56
EUR
92326.
92327.
92325.
Antibody labelling kits, Mix-n-Stain™ DNP
Systémy pro spojování a značení proteinů
Mix-n-Stain™ DNP antibody labelling kits utilise a technology that allows rapid antibody labelling with dinitrophenol without a purification step.
- The labelling procedure tolerates many common buffer components and antibody stabilisers