Tlačiť
Polycytidylic acid - agarose
Dodávateľ: Merck
P9827-1G P9827-250MG
SIALP9827-1GEA 1822.7 EUR
SIALP9827-1G SIALP9827-250MG
Polycytidylic acid - agarose
Gely elektroforetické Agaróza a Akrylamid
Polycytidylic acid-agarose is used in protein chromatography, affinity chromatography and speciality resins.
Polycytidylic acid is used to study the preferential induction of apoptosis in the rainbow trout macrophage cell line and also to develop two analytical methods for human secretory-type ribonuclease.