Tlačiť
Density hydrometers, S50
Dodávateľ: Amarell
H811108 H811100 H811102 H811104 H811106 H811110 H811112 H811114 H811116 H811118 H811120 H811122 H811124 H811126 H811128 H811130 H811132 H811134 H811136 H811138 H811140 H811142 H811144 H811146 H811148 H811150 H811152 H811154
AMARH811108EA 62.58 EUR
AMARH811108 AMARH811100 AMARH811102 AMARH811104 AMARH811106 AMARH811110 AMARH811112 AMARH811114 AMARH811116 AMARH811118 AMARH811120 AMARH811122 AMARH811124 AMARH811126 AMARH811128 AMARH811130 AMARH811132 AMARH811134 AMARH811136 AMARH811138 AMARH811140 AMARH811142 AMARH811144 AMARH811146 AMARH811148 AMARH811150 AMARH811152 AMARH811154
Density hydrometers, S50
Hydrometry
Hydrometers in a wide range of measuring scales.
  • Shot weighted body
  • Paper scale with colour code and type designation
Suitable for government verification.
Certifikácie: DIN 12791
Balenie: In a transparent plastic case.