Tlačiť
Transfection reagent, in vivo PolyMag™
Dodávateľ: OZ BIOSCIENCES
IV-PN30500 IV-PN31000 IV-TK30210 IV-KC30210
OZBIIV-PN30500EA 402.33 EUR
OZBIIV-PN30500 OZBIIV-PN31000 OZBIIV-TK30210 OZBIIV-KC30210
Transfection reagent, in vivo PolyMag™
Reagencie transfekční
In vivo PolyMag™ transfection reagent has been designed for in vivo targeted transfection of various types of nucleic acids, such as DNA, RNA, oligonucleotides, using magnetic nanoparticles coated with specific polymers. DNA complexes can be easily administrated through various injection routes, such as systemic administration (intravenous, intra-artery) or local administration (intratumoral, intracerebroventricular).
  • Increased transfection efficiency
  • Targeted process (magnetically-driven)
  • Reduction of the systemic dissemination of vectors during injection
  • Reduction of the vector doses
  • Work under non permissive conditions (hypothermia, physiological flow conditions)
  • Penetration of the vector into tissues
  • MInimised toxicity
  • Universal suitable for all nucleic acids
Store at +4 °C.
Informácie o príslušenstve: Needs to be used with a magnetic plate, available as a separate accessory or as part of a starter kit.
Informace o dodávce: Shipped at room temperature.