Tlačiť
ESD safety ankle boots, lace-up, Wellmax, Reaction XXT
Dodávateľ: ELTEN
769701/36 769701/37 769701/38 769701/39 769701/40 769701/41 769701/42 769701/43 769701/44 769701/45 769701/46 769701/47 769701/48
ELTN769701/36EA 120.34 EUR
ELTN769701/36 ELTN769701/37 ELTN769701/38 ELTN769701/39 ELTN769701/40 ELTN769701/41 ELTN769701/42 ELTN769701/43 ELTN769701/44 ELTN769701/45 ELTN769701/46 ELTN769701/47 ELTN769701/48
ESD safety ankle boots, lace-up, Wellmax, Reaction XXT
Obuv
Metal-free ESD safety ankle boots made from waxy cowhide leather, full, padded bellows tongue plus a dual-density PU/PU sole (Wellmaxx Trainers).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), black
  • Anatomically-shaped insole and ankle support for improved stability
  • Cushioned midsole made of Infinergy® (by BASF) rebounds back extremely well when under pressure, giving energy back to the wearer
  • Abrasion-proof scuff cap
  • Metal-free, puncture resistant midsole
  • Composite toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifikácie: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.