Tlačiť
ESD safety shoes, sandals, Velcro® fastening, Nova, Maja Easy S1P
Dodávateľ: ELTEN
74771/35 74771/36 74771/37 74771/38 74771/39 74771/40 74771/41 74771/42
ELTN74771/35EA 95.87 EUR
ELTN74771/35 ELTN74771/36 ELTN74771/37 ELTN74771/38 ELTN74771/39 ELTN74771/40 ELTN74771/41 ELTN74771/42
ESD safety shoes, sandals, Velcro® fastening, Nova, Maja Easy S1P
Obuv
Leather-free, lightweight ESD safety sandal made from microfibre/textile upper, a padded tongue plus collar and a slip resistant, dual-density PU/PU sole (Nova).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (Ladies ESD)
  • Soft fleece ESD insole
  • Puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in dry work areas such as industry, storage, transport, construction etc.
Colour: Black
Certifikácie: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC.