Tlačiť
Test sieves, stainless steel
Dodávateľ: ENDECOTTS
008SIW.071 008SIW.075 008SIW.080 008SIW.090 008SIW.100 008SIW.106 008SIW.125 008SIW.140 008SIW.150 008SIW.160 008SIW.180 008SIW.200 008SIW.212 008SIW.224 008SIW.250 008SIW.280 008SIW.300 008SIW.315 008SIW.355 008SIW.400 008SIW.425 008SIW.450 008SIW.500 008SIW.560 008SIW.600 008SIW.630 008SIW.800 008SIW.850 008SIW.900 008SIW.045 008SIW1.00 008SIW1.12 008SIW1.18 008SIW1.25 008SIW1.40 008SIW1.60 008SIW1.70 008SIW1.80 008SIW10.0 008SIW100 008SIW106 008SIW11.2 008SIW112 008SIW125 008SIW13.2 008SIW14.0 008SIW16.0 008SIW18.0 008SIW19.0 008SIW2.00 008SIW2.24 008SIW2.36 008SIW2.50 008SIW2.80 008SIW20.0 008SIW22.4 008SIW25.0 008SIW26.5 008SIW28.0 008SIW3.15 008SIW3.35 008SIW3.55 008SIW31.5 008SIW35.5 008SIW37.5 008SIW4.00 008SIW4.50 008SIW4.75 008SIW40.0 008SIW45.0 008SIW5.00 008SIW5.60 008SIW50.0 008SIW53.0 008SIW56.0 008SIW6.30 008SIW6.70 008SIW63.0 008SIW7.10 008SIW75.0 008SIW8.00 008SIW80.0 008SIW9.00 008SIW9.50 008SIW90.0 008SIW12.5 008SIW.112 008SIW.710 008SIW71.0 008SIW.020 008SIW.025 008SIW.032 008SIW.036 008SIW.038 008SIW.040 008SIW.050 008SIW.053 008SIW.056 008SIW.063
ENDE008SIW.071EA 251.5 EUR
ENDE008SIW.071 ENDE008SIW.075 ENDE008SIW.080 ENDE008SIW.090 ENDE008SIW.100 ENDE008SIW.106 ENDE008SIW.125 ENDE008SIW.140 ENDE008SIW.150 ENDE008SIW.160 ENDE008SIW.180 ENDE008SIW.200 ENDE008SIW.212 ENDE008SIW.224 ENDE008SIW.250 ENDE008SIW.280 ENDE008SIW.300 ENDE008SIW.315 ENDE008SIW.355 ENDE008SIW.400 ENDE008SIW.425 ENDE008SIW.450 ENDE008SIW.500 ENDE008SIW.560 ENDE008SIW.600 ENDE008SIW.630 ENDE008SIW.800 ENDE008SIW.850 ENDE008SIW.900 ENDE008SIW.045 ENDE008SIW1.00 ENDE008SIW1.12 ENDE008SIW1.18 ENDE008SIW1.25 ENDE008SIW1.40 ENDE008SIW1.60 ENDE008SIW1.70 ENDE008SIW1.80 ENDE008SIW10.0 ENDE008SIW100 ENDE008SIW106 ENDE008SIW11.2 ENDE008SIW112 ENDE008SIW125 ENDE008SIW13.2 ENDE008SIW14.0 ENDE008SIW16.0 ENDE008SIW18.0 ENDE008SIW19.0 ENDE008SIW2.00 ENDE008SIW2.24 ENDE008SIW2.36 ENDE008SIW2.50 ENDE008SIW2.80 ENDE008SIW20.0 ENDE008SIW22.4 ENDE008SIW25.0 ENDE008SIW26.5 ENDE008SIW28.0 ENDE008SIW3.15 ENDE008SIW3.35 ENDE008SIW3.55 ENDE008SIW31.5 ENDE008SIW35.5 ENDE008SIW37.5 ENDE008SIW4.00 ENDE008SIW4.50 ENDE008SIW4.75 ENDE008SIW40.0 ENDE008SIW45.0 ENDE008SIW5.00 ENDE008SIW5.60 ENDE008SIW50.0 ENDE008SIW53.0 ENDE008SIW56.0 ENDE008SIW6.30 ENDE008SIW6.70 ENDE008SIW63.0 ENDE008SIW7.10 ENDE008SIW75.0 ENDE008SIW8.00 ENDE008SIW80.0 ENDE008SIW9.00 ENDE008SIW9.50 ENDE008SIW90.0 ENDE008SIW12.5 ENDE008SIW.112 ENDE008SIW.710 ENDE008SIW71.0 ENDE008SIW.020 ENDE008SIW.025 ENDE008SIW.032 ENDE008SIW.036 ENDE008SIW.038 ENDE008SIW.040 ENDE008SIW.050 ENDE008SIW.053 ENDE008SIW.056 ENDE008SIW.063
Test sieves, stainless steel
Síta Síta
These woven wire mesh sieves are the most widely used test sieves for all types of laboratory sampling and particle size analysis.
  • Precision frame ensures consistent nestability
  • Precise aperture
  • Natural fillet for free flowing of sample
  • Totally sealed, no crevice to lose material
  • Evenly tensioned mesh ensures accurate analysis
  • Safe edge with big radius makes it comfortable to handle
  • Serial number ensures full traceability
These sieves have full height.
Certifikácie: ISO 3310-1/BS410.