Tlačiť

Ethyl nicotinate, Sigma-Aldrich®

Dodávateľ: Merck

Synonymá: 3-Pyridinecarboxylic acid ethyl ester, Ethyl 3-pyridinecarboxylate, Pyridine-3-carboxylic acid ethyl ester, 3-Picolinic acid ethyl ester, Nicotinic acid ethyl ester

M-Clarity™ quality level = MQ200

E40609-100G E40609-500G
SIALE40609-100GEA 51.74 EUR
SIALE40609-100G SIALE40609-500G
Ethyl nicotinate, Sigma-Aldrich®
Ethyl nicotinate
Vzorec: C₈H₉NO₂
Mw: 151.16
Teplota varu: 223
Hustota: 1.115 g/cm³ (20 °C)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00006389
Číslo CAS: 614-18-6
EINECS: 210-370-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR