Tlačiť

2-Amino-6-chloropurine, Sigma-Aldrich®

Dodávateľ: Merck

Synonymá: 6-Chloroguanine, 6-Chloro-7H-purin-2-ylamine

M-Clarity™ quality level = MQ100

342300-1G 342300-5G
SIAL342300-1GEA 56.88 EUR
SIAL342300-1G SIAL342300-5G
2-Amino-6-chloropurine, Sigma-Aldrich®
2-Amino-6-chloropurine
Vzorec: C₅H₄ClN₅
Mw: 169.57
Teplota tavenia: 300
Číslo MDL: MFCD00075252
Číslo CAS: 10310-21-1
EINECS: 233-686-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR