Tlačiť

Platinum(II) chloride, Sigma-Aldrich®

Dodávateľ: Merck

Synonymá: Platinum dichloride

M-Clarity™ quality level = MQ100

206091-1G 206091-250MG 206091-5G
SIAL206091-1GEA 188.13 EUR
SIAL206091-1G SIAL206091-250MG SIAL206091-5G
Platinum(II) chloride, Sigma-Aldrich®
Platinum(II) chloride
Vzorec: Cl₂Pt
Mw: 265.99
Teplota tavenia: 581
Hustota: 6.05 g/cm³ (20 °C)
Číslo MDL: MFCD00011181
Číslo CAS: 10025-65-7
EINECS: 233-034-1
OSN: 3260
ADR: 8 III
Merck Index: 12,07687

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR