Tlačiť
Phosphate buffered saline (PBS), Sigma-Aldrich®
Dodávateľ: Merck
P3563-10PAK P3563-5X10PAK
SIALP3563-10PAKEA 86.64 EUR
SIALP3563-10PAK SIALP3563-5X10PAK
Phosphate buffered saline (PBS), Sigma-Aldrich®
Pufry Pufry pro tkáňové kultury
Contents of one pouch, when dissolved in one liter of distilled or deionised water, will yield 0,01 M phosphate buffered saline (NaCl 0,138 M; KCl - 0,0027 M); TWEEN® 20 - 0,05%, pH 7,4, at 25 °C.