Tlačiť
Industrial scales, FG series
Dodávateľ: AnD Instruments Ltd (A&D)
FG-60KBM-EC FG-150KAL FG-150KAL-EC FG-30KBM FG-30KBM-EC FG-60KAL FG-60KAL-EC FG-60KBM
ANDAFG-60KBM-ECEA 0 EUR
ANDAFG-60KBM-EC ANDAFG-150KAL ANDAFG-150KAL-EC ANDAFG-30KBM ANDAFG-30KBM-EC ANDAFG-60KAL ANDAFG-60KAL-EC ANDAFG-60KBM
Industrial scales, FG series
Váhy
The FG series of scales combine fast response time and rock-solid durability in an easy to use package.
  • Full digital calibration
  • Auto power off and counting function
  • Status indicators (stable, zero, and net)
  • High-speed weighing with one-second stabilisation
  • Washable and hygienic stainless steel weighing pan in two sizes
  • Three selectable weighing resolutions
  • Comparator output (FG-OP-23/FG-OP-24)
Display unit can be either on a column (FG-KAM/KAL) or attached directly to the platform (FG-KBM). The scale can be powered by AC adaptor or dry cell battery. FG scales are availabe with OIML-EC approval and all models feature a IP65 dust and water protected loadcell.
Additional weighing modes include parts counting and HI/OK/LOW comparator function for checkweighing.