Germanium(IV) ethoxide ≥99.995% (metals basis)
Dodávateľ: Thermo Fisher Scientific
|
Varování
|
Purity: 99.995%
CAS No.: 14165-55-0
Molecular Formula: C8H20GeO4
Formula Weight: 252.84
Flash Point: 49°(121°F)
Physical Form: Liquid
Density: 1.14
Refractive Index: 1.4070
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Alcohol-like
Melting Point: -72°
Boiling Point: 185.5°
UN No.: 1993
MDL No.: MFCD00015121
Preparation of GeO2 by CVD; intermediate in sol-gel synthesis
Vzorec:
C₈H₂₀GeO₄ Teplota varu: 185,5 °C Teplota tavenia: –72 °C Hustota: 1,14 Teplota vzplanutia: 49 °C (121 °F) Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00015121 Číslo CAS: 14165-55-0 EINECS: 238-007-8 OSN: 1993 ADR: 3,III |