Tlačiť
Volumetric flasks, class A, blue markings, BLAUBRAND®
Dodávateľ: BRAND
937048 937045 937047
BRND937048EA 60.97 EUR
BRND937048 BRND937045 BRND937047
Volumetric flasks, class A, blue markings, BLAUBRAND®
Baňky Baňky odměrné
Borosilicate glass 3,3, with beaded rim.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
* Wide neck
Certifikácie: DIN EN ISO 1042
Objednávacie informácie: Supplied with an individual certificate.