L(-)-Glutathion (redukovaná forma) ≥97%
Dodávateľ: Thermo Fisher Scientific
Synonymá:
L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
A18014.06
A18014.14
A18014.22
A18014.06EA
65.29
EUR
A18014.06
A18014.14
A18014.22
L(-)-Glutathion (redukovaná forma) ≥97%
L(-)-Glutathion (redukovaná forma)
Purity: 97%
CAS No.: 70-18-8
Molecular Formula: C10H17N3O6S
Formula Weight: 307.33
Storage Temperature: 2°C to 8°C
Melting Point: ca 193° dec.
Beilstein Reference: 1729812
Merck Reference: 14,4475
Optical Rotation: -17° (c=2 in water)
MDL No.: MFCD00065939
Vzorec:
C₁₀H₁₇N₃O₆S Mw: 307.33 Teplota tavenia: ∼ 193 °C (rozkl.) Hustota: 1.475 g/cm³ (20 °C) Skladovacia teplota: Refrigerator |
Číslo MDL:
MFCD00065939 Číslo CAS: 70-18-8 EINECS: 200-725-4 Merck Index: 13,04488 |