Tlačiť

Silver methanesulphonate

Dodávateľ: Thermo Fisher Scientific
Nebezpečí

Synonymá: Methanesulphonic acid silver salt

18641.09 18641.18
18641.09EA 162.45 EUR
18641.09 18641.18
Silver methanesulphonate
Silver methanesulphonate

CAS No.: 2386-52-9 Molecular Formula: CH3AgO3S Formula Weight: 202.97 Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to grey Odour: Odourless UN No.: 3077 MDL No.: MFCD00064795

Vzorec: CH₃AgO₃S
Teplota tavenia: 263 °C
Skladovacia teplota: Ambient
Číslo MDL: MFCD00064795
Číslo CAS: 2386-52-9
EINECS: 219-199-2
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR