Tlačiť

Methanesulphonic anhydride, Sigma-Aldrich®

Dodávateľ: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

269190-25G 269190-5G
SIAL269190-25GEA 122.97 EUR
SIAL269190-25G SIAL269190-5G
Methanesulphonic anhydride, Sigma-Aldrich®
Methanesulphonic anhydride
Vzorec: C₂H₆O₅S₂
Mw: 174.2
Teplota tavenia: 63…66 °C
Skladovacia teplota: Refrigerator
Číslo MDL: MFCD00007556
Číslo CAS: 7143-01-3
EINECS: 230-442-1
OSN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR