Tlačiť
Safety ankle boots, lace-up, jo_Twist Boa Mid, Jori
Dodávateľ: ELTEN
16761/43 16761/40 16761/45 16761/38 16761/39 16761/47 16761/41 16761/46 16761/44 16761/48 16761/42
ELTN16761/43EA 109.51 EUR
ELTN16761/43 ELTN16761/40 ELTN16761/45 ELTN16761/38 ELTN16761/39 ELTN16761/47 ELTN16761/41 ELTN16761/46 ELTN16761/44 ELTN16761/48 ELTN16761/42
Safety ankle boots, lace-up, jo_Twist Boa Mid, Jori
Obuv
Anti-static safety ankle boot made from nubuck leather and textile material, a full padded bellows tongue, plus a dual-density PU/PU sole (oil, fuel and slip resistant, heat resistant up to approx. 130 °C).
  • Climate-regulating, breathable textile lining
  • Lace- and buckle-free Boa® system can be opened and closed with just one hand
  • Full length inlay sole
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifikácie: EN ISO 20345; S1P, SRC.