Tlačiť

Undecanoic acid, Sigma-Aldrich®

Dodávateľ: Merck

Synonymá: Hendecanoic acid, Undecylic acid

M-Clarity™ quality level = MQ100

171476-100G 171476-25G
SIAL171476-100GEA 172.08 EUR
SIAL171476-100G SIAL171476-25G
Undecanoic acid, Sigma-Aldrich®
Undecanoic acid
Vzorec: C₁₁H₂₂O₂
Mw: 186.29
Teplota varu: 228
Teplota tavenia: 30
Hustota: 0.89 g/cm³ (20 °C)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00002730
Číslo CAS: 112-37-8
EINECS: 203-964-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR