Octanoyl chloride ≥99%
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
L03526.AE
L03526.AP
L03526.AEEA
33.17
EUR
L03526.AE
L03526.AP
Octanoyl chloride ≥99%
Octanoyl chloride
Purity: 99%
CAS No.: 111-64-8
Molecular Formula: CH3(CH2)6COCl
Formula Weight: 162.66
Flash Point: 80°(176°F)
Refractive Index: 1.4350
Odour: Pungent
Melting Point: -63°
Boiling Point: 195°
Beilstein Reference: 635917
UN No.: 3265
MDL No.: MFCD00000766
Vzorec:
C₈H₁₅ClO Teplota varu: 195 °C Teplota tavenia: –63 °C Hustota: 0,942 Teplota vzplanutia: 80 °C (176 °F) Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00000766 Číslo CAS: 111-64-8 EINECS: 203-891-6 OSN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...