Tlačiť

Hydrazinium hydroxid ≥98% N₂H₄ 64-65 %, Reagent Grade, Sigma-Aldrich®

Dodávateľ: Merck
Nebezpečí

Synonymá: Hydrazine hydrate, Hydrazine monohydrate

M-Clarity™ quality level = MQ100

207942-2KG 207942-500G 207942-100G 207942-5G
SIAL207942-2KGEA 222.31 EUR
SIAL207942-2KG SIAL207942-500G SIAL207942-100G SIAL207942-5G
Hydrazinium hydroxid ≥98% N₂H₄ 64-65 %, Reagent Grade, Sigma-Aldrich®
Hydrazinium hydroxid
Vzorec: NH₂NH₂·H₂O
Mw: 50.06
Teplota varu: 117.2
Teplota tavenia: –51.7 °C
Hustota: 1.03 g/cm³ (20 °C)
Teplota vzplanutia: 75
Číslo MDL: MFCD00149931
Číslo CAS: 7803-57-8
EINECS: 206-114-9
OSN: 2030
ADR: 8 II
Merck Index: 13,04790

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR