(±)-α-Methylbenzylamine ≥98%
Dodávateľ: abcr
|
Nebezpečí
|
Synonymá:
DL-1-Phenylethylamine, (±)-1-Phenylethylamine, (±)-α-Phenylethylamine
AB118493.0100
AB118493.0500
ABCRAB118493.0100EA
0
EUR
ABCRAB118493.0100
ABCRAB118493.0500
(±)-α-Methylbenzylamine ≥98%
(±)-α-Methylbenzylamine
Purity: 98+%
CAS No.: 618-36-0
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 79°(174°F)
Refractive Index: 1.5270
Odour: Odourless
Boiling Point: 69-70°/10mm
Beilstein Reference: 636127
Merck Reference: 14,6026
UN No.: 2735
MDL No.: MFCD00008069
Vzorec:
C₆H₅CH(NH₂)CH₃ Teplota varu: 69…70 °C (10 mmHg) Teplota tavenia: –65 °C Hustota: 0,945 Teplota vzplanutia: 79 °C (174 °F) Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00008069 Číslo CAS: 618-36-0 EINECS: 210-545-8 OSN: 2735 ADR: 8,III Merck Index: 14,6026 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...