Tlačiť
Bulk media for HPLC, NUCLEODUR® C18 ec
Dodávateľ: MACHEREY-NAGEL
713550.0100 713601.0100 713611.0100 713621.0100 713631.0100 713618.0100
MANA713550.0100EA 1032.89 EUR
MANA713550.0100 MANA713601.0100 MANA713611.0100 MANA713621.0100 MANA713631.0100 MANA713618.0100
Bulk media for HPLC, NUCLEODUR® C18 ec
Chromatografii, Sorbenty HPLC sorbenty
High purity silica phase for HPLC.
  • Totally spherical high purity silica
  • Pressure stable up to 600 bar
  • Pore volume 0,9 ml/g
  • Density 0,47 g/ml
  • Carbon content 17,5%
  • pH stability 1 to 9
  • Reliable standard RP phase for up-scaling to preparative scale
NUCLEODUR® C18 ec with its medium density octadecyl modification and an exhaustive endcapping is recommended for separartions in a wide range of application areas such like basic, neutral or acidic substances, aldehydes and ketones as well as phenolic compounds.