Tlačiť
Erlenmeyer flasks, disposable, with vented screw cap, Nalgene®
Dodávateľ: Thermo Fisher Scientific
4116-2000 4116-1000 4116-0500 4116-0250 4116-0125 4115-2000 4115-1000 4115-0500 4115-0250 4115-0125
214-7215EA 137.99 EUR
214-7215 214-7214 214-7213 214-7212 214-7211 214-7210 214-7209 214-7208 214-7207 214-7206
Erlenmeyer flasks, disposable, with vented screw cap, Nalgene®
Baňky Baňky dle Erlenmeyera
PETG, transparent with blue HDPE vented screw cap, flat or baffled bottom.
  • Sterile disposable flasks reduce the chance of cross contamination
  • Moulded-in graduations
  • Flasks offer a five year shelf life, a 10⁻⁶ SAL, are non pyrogenic and non cytotoxic
Ideal for shakers and suspension cell culture, media preparation or storage. HDPE closure has a hydrophobic 0,2 µm PTFE membrane that allows sterile gas exchange.
Balenie: Individually packaged for easy storage and handling.