Tlačiť
Cryoboxes, 136×136 mm
Dodávateľ: TENAK
TE22525 TE22502 TE22500 TE22736 TE22540 TE22704 TE22686 TE22684 TE22683 TE22710 TE22492 TE22494 TE22496 TE22498 TE22669 TE22675 TE22679 TE22690 TE22702
479-0703EA 3.65 EUR
479-0703 479-0700 479-0698 479-0724 479-0708 479-0718 479-0716 479-0714 479-0713 TENATE22710 TENATE22492 TENATE22494 TENATE22496 TENATE22498 TENATE22669 TENATE22675 TENATE22679 TENATE22690 TENATE22702
Cryoboxes, 136×136 mm
Krabice Kryoboxy
Cryo resistant fibreboard/cardboard.
  • Water repellent for longer durability
  • Various combinations possible with different dividers (to be ordered separately)
  • Available in different heights and colours
For storage of cryovials at low temperatures.
*Height stated is the interior height.