Tlačiť
Ac-WEHD-AFC
Dodávateľ: Enzo Life Sciences
ALX260117M005 ALX260117M010
ENZOALX260117M005EA 361.41 EUR
ENZOALX260117M005 ENZOALX260117M010
Ac-WEHD-AFC
Proteiny
Substrate for caspase-1, -4- and -5
Fluorogenic substrate for caspase-1 and -5. Also a substrate for caspase-4.