Tlačiť
Anti-PLK1 Mouse Monoclonal Antibody
Dodávateľ: ENZO LIFE SCIENCES
ADIKAMCC105D ADIKAMCC105F
ENZOADIKAMCC105DEA 314.48 EUR
ENZOADIKAMCC105D ENZOADIKAMCC105F
Anti-PLK1 Mouse Monoclonal Antibody
Protilátky
The polo-like kinases (PLKs) function in cell-cycle progression and in multiple stages of mitosis. PLK1 is activated by phosphorylation of Thr210 at the G2/M phase boundary. Active PLK1 participates in mitotic entry through activation of Cdc25C and nuclear import of cyclin B1, thereby activating the Cdc2/cyclin B kinase.
  • For Western Blot
Recognizes human, mouse (weak) and rat Plk1. Detects a band of ~62kDa by Western blot.
Type: Primary
Antigen: PLK1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human