Tlačiť
Round cover glasses, #1,5
Dodávateľ: MARIENFELD
0117580 0117520 0117650 0117640 0117500 0117530
630-2200EA 356.6 EUR
630-2200 630-2190 630-2213 630-2204 630-2189 MARI0117530
Round cover glasses, #1,5
Sklíčka krycí
Cover slides recommended for objectives with high numerical aperture and resolution.
  • Borosilicate glass D 263™ M of hydrolytic class 1
Suitable for fluorescence microscopy.