Tlačiť
Sorbents, LiChrosorb®
Dodávateľ: Merck
1.09309.0010 1.09318.0010 1.09332.0010 1.09333.0010 1.09334.0010
1.09309.0010EA 231.85 EUR
1.09309.0010 1.09318.0010 1.09332.0010 1.09333.0010 1.09334.0010
Sorbents, LiChrosorb®
Chromatografii, Sorbenty HPLC sorbenty
LiChrosorb® is an irregular shaped silica sorbent. LiChrosorb® packing materials offer the compete programme of non-polar derivatives (RP-8, RP-18, RP-select B) and polar derivatives (Si 60 and Si 100).