Tlačiť
Multi-channel pipettes, mechanical, variable volume, Reference® 2 (General Lab Product)
Dodávateľ: EPPENDORF
4926000.018 4926000.026 4926000.034 4926000.042 4926000.050 4926000.069
613-5843EA 1185.72 EUR
613-5843 613-5844 613-5845 613-5846 613-5847 613-5848
Multi-channel pipettes, mechanical, variable volume, Reference® 2 (General Lab Product)
Pipety Micropipettes
The robust Reference® 2 is the successor to the Reference® pipette and features a new design, reduced weight and unique one-button operation.
  • One-button operation and spring loaded tip cone for fast and ergonomic handling with reduced operating effort
  • Quick tip ejection with active aerosol reduction; haptic feedback prevents accidential tip ejection
  • Spring-loaded tip cone can be switched on/off optionally and improves reproducibility and flexibility
  • Channel indicator guarantees a continuously identical pipette alignment during the work process
  • Serial number is printed on multiple components of the pipette for heightened traceability
  • Embedded RFID chip containing all relevant data ensures complete traceability
  • Optional TrackIT software allows simple identification and documentation and provides ability to add own data (e.g. calibration time and results, location)
  • Fully autoclavable
Thanks to its reliable and accurate operation, the pipette is ideal for use with precious liquids or in any application requiring extremely high levels of accuracy.
Designed and constructed for general laboratory and research applications in Life Science. Typical applications include: Food & beverages, forensic, cosmetics, agriculture, animal studies, chemical analytics, research applications at medical and academic facilities.
* Works with Windows 7/8 and Vista.
Certifikácie: Without CE marking.