Tlačiť
Selenium standards, VHG
Dodávateľ: LGC Standards PROMOCHEM
VHG-SPSE4-100 VHG-SPSE6-100 VHG-SPSE4-50 VHG-SPSE6-50
PROOVHG-SPSE4-100EA 0 EUR
PROOVHG-SPSE4-100 PROOVHG-SPSE6-100 PROOVHG-SPSE4-50 PROOVHG-SPSE6-50
Selenium standards, VHG
Standardy Standardy kationické Standardy - selen
This standard solution is intended for use as a calibration and reference standard for plasma emission spectroscopy (ICP or DCP), inductively coupled plasma mass spectroscopy (ICP-MS), atomic absorption spectroscopy (AA or GFAA) or techniques using other modes of elemental detection and an appropriate separation method.
Certifikácie: Accredited to ISO 9001 and ISO/IEC 17025. Traceable to NIST and SRM.