Tlačiť
Meat extract, Sigma-Aldrich®
Dodávateľ: Merck
70164-100G 70164-500G
SIAL70164-100GEA 0 EUR
SIAL70164-100G SIAL70164-500G
Meat extract, Sigma-Aldrich®
Média mikrobiologická Mikrobiologické přísady a doplňky
Meat extract is manufactured from meat with low fat and sinew content and can be considered as complementing the nutritive properties of peptone by contributing minerals, phosphates, energy sources and those essential factors missing from peptone.
  • Coagulable protein, none detected
  • Nitrogen content: 11,5 to 12,5% total nitrogen (N) and 3,5 to 4,5% amino nitrogen
  • Ignition residue: ≤18%