Tlačiť
Cobalt standard solution, for ICP, in 5% HNO₃, VHG
Dodávateľ: LGC Standards PROMOCHEM
VHG-PCON-100 VHG-PCON-50 VHG-PCON-500 VHG-TCON-100 VHG-TCON-500
PROOVHG-PCON-100EA 65.78 EUR
PROOVHG-PCON-100 PROOVHG-PCON-50 PROOVHG-PCON-500 PROOVHG-TCON-100 PROOVHG-TCON-500
Cobalt standard solution, for ICP, in 5% HNO₃, VHG
Standardy Standardy kationické Standardy - kobalt
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifikácie: Traceable to NIST, SRM.