Tlačiť
Molybdenum standard solution, for ICP, in 5% HNO₃, VHG
Dodávateľ: LGC Standards PROMOCHEM
VHG-PMONF-100 VHG-PMONF-50 VHG-PMONF-500 VHG-TMONF-100 VHG-TMONF-500
PROOVHG-PMONF-100EA 65.78 EUR
PROOVHG-PMONF-100 PROOVHG-PMONF-50 PROOVHG-PMONF-500 PROOVHG-TMONF-100 PROOVHG-TMONF-500
Molybdenum standard solution, for ICP, in 5% HNO₃, VHG
Standardy Standardy kationické Standardy - molybden
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifikácie: Traceable to NIST, SRM.