Tlačiť
Group lockout box, wall-mountable, SAFETY REDBOX®
Dodávateľ: BRADY
145579 145580
118-0529EA 146.41 EUR
118-0529 BRDY145580
Group lockout box, wall-mountable, SAFETY REDBOX®
Zámky bezpečnostní
This wall-mountable and portable lock box features a quick release internal slide button that allows the lockbox to be carried to the point of need.
  • Box can hold upto 12 padlocks
  • Made of heavy-duty, powder coated steel for rust-resistance and durability under most environmental conditions
  • Use one lock on each energy control point and place the keys in the lock box; each worker then places his own lock on the box to prevent access
  • Each employee retains exclusive control, as required by OSHA, by placing his own lock on the lock box containing the keys to the job locks
  • As long as any one worker's lock remains on the lock box, the keys to the job locks contained inside cannot be accessed