Tlačiť
Decontamination solution, DNA AWAY®
Dodávateľ: Thermo Fisher Scientific
7010 7012-11
732-2353EA 20.9 EUR
732-2353 732-4436
Decontamination solution, DNA AWAY®
Prostředky čistící Dekontaminanty
Ready-to-use solution for eliminating DNA contamination. For cleaning work surfaces, instruments, pipettes, thermal cyclers and more.
  • Suitable for PCR applications, preventing false-positive PCR results
  • Easy cleaning of various types of surfaces