Test kits, photometric, Nitrate, LCK cuvette tests
Dodávateľ: Hach
LCK340
LCK339
705-3053EA
193.74
EUR
705-3053
705-3052
Test kits, photometric, Nitrate, LCK cuvette tests
Kity pro environmentální testy
Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C
Certifikácie: ISO 7890-1-2-1986, DIN 38405 D9-2.