Tlačiť
Syringe filters, Captiva
Dodávateľ: AGILENT HEWLETT PACKARD
5190-5126 5190-5127 5190-5128 5190-5129 5190-5132 5190-5133 5190-5134 5190-5135
HEWL5190-5126EA 395.11 EUR
HEWL5190-5126 HEWL5190-5127 HEWL5190-5128 HEWL5190-5129 HEWL5190-5132 HEWL5190-5133 HEWL5190-5134 HEWL5190-5135
Syringe filters, Captiva
Filtry Filtry stříkačkové
These syringe filters are designed for filtering samples prior to HPLC, UHPLC, and LC/MS analysis. This can improve both system performance and analytical quality.
  • Available in a wide range of sizes, formats, and membranes
Every syringe filter is manufactured using the highest-grade housing and membrane materials for excellent flow, maximum sample loading capacity, and to ensure sample integrity.