Tlačiť
Biohazard bag holders, Poxygrid® wire, SP Bel-Art
Dodávateľ: Bel-Art Products, a Part of SP
H131870000 H131880000 H131890000
129-1022EA 0 EUR
129-1022 129-1023 129-1024
Biohazard bag holders, Poxygrid® wire, SP Bel-Art
Stojany Držáky sáčků
These epoxy-coated, wire framed bag holders keep biohazard bags open and ready for use. The user just has to place a biohazard bag into the sturdy wire holder and stretch it over the top ring to secure it in place. A separate hooked ring rests over the top to keep the bag in place. Circular shape keeps bags wide open for easy access.
  • Made from bright biohazard orange-coloured, epoxy-coated wire