Tlačiť
Eyewash stations with two bottles
Dodávateľ: Plum Safety
4650 4694
118-1702EA 78.62 EUR
118-1702 118-1705
Eyewash stations with two bottles
Pomůcky první pomoci Sprchy na výplach očí
Available as either an open station or closed and dust-proof wall box containing 2×500 ml plum eyewash bottles.
  • Can be easily installed close to the workplace
  • Open station with mirror and pictogram, wall box with mirror and eye wash sign
  • Open station not suitable for very dusty or dirty workplaces
Rinsing time: ~10 minutes
W×D×H open station: 230×80×290 mm
W×D×H boxed station: 227×110×265 mm