Tlačiť
REDExtract-N-Amp™ PCR ReadyMix™
Dodávateľ: Merck
R4775-125ML R4775-12ML
SIALR4775-125MLEA 0 EUR
SIALR4775-125ML SIALR4775-12ML
REDExtract-N-Amp™ PCR ReadyMix™
Činidla pro nukleové kyseliny End-point PCR Enzymes and Kits
PCR ReadyMix™ is intended for use with Extract-N-Amp™ Plant PCR kit and Extract-N-Amp™ Tissue PCR kit. All Extract-N-Amp kits include a PCR ReadyMix™ sufficient for one PCR reaction per extraction. However, where additional PCR reactions are required, supplemental PCR ReadyMix™ may be needed. The REDExtract-N-Amp™ PCR ReadyMix contains a inert tracking dye that allows for convenient direct loading of PCR reactions onto agarose gels for analysis.