Tlačiť
Vložky do rukavic, HyFlex® 48-105
Dodávateľ: Ansell
48-105/6 48-105/7 48-105/8 48-105/9 48-105/10 48-105/11
112-3782EA 41.72 EUR
112-3782 112-3783 112-3784 112-3785 112-3786 112-3787
Vložky do rukavic, HyFlex® 48-105
Rukavice
Knitted nylon gloves with PU coated fingertips.
  • Optimum protection for light tasks, high level of comfort
  • Good freedom of movement for precise handling of small objects in dry conditions
  • Elasticated nylon liner for exceptionally close fit
  • Excellent alternative to light leather gloves
Ideal for light duty applications, such as assembling, inspecting and packing small components.
Colour: White
EN 388 (003X), Cat. II
Certifikácie: EN 388:2016 (003XA); Cat. II