Tlačiť
Beakers, low form, PFA, Corning®
Dodávateľ: Corning
1003P-100 1003P-1L 1003P-250 1003P-50 1003P-500 1003P-25
CORN1003P-100EA 619.33 EUR
CORN1003P-100 CORN1003P-1L CORN1003P-250 CORN1003P-50 CORN1003P-500 CORN1003P-25
Beakers, low form, PFA, Corning®
Kádinky
Made of PFA, translucent.
  • Excellent chemical resistance
  • Good temperature resistance
With a raised graduation scale.