Tlačiť
Single-channel pipettes, electronic, with Bluetooth, E1-ClipTip™
Dodávateľ: Thermo Fisher Scientific
4670000BT 4670020BT 4670030BT 4670040BT
613-5748EA 1125.95 EUR
613-5748 613-5749 613-5750 613-5751
Single-channel pipettes, electronic, with Bluetooth, E1-ClipTip™
Pipety Micropipettes
These pipettes are ideal for repetitive pipetting tasks with microcentrifuge or test tubes. Electronic tip ejection and index finger pipetting action allow the thumb to relax during pipetting.
  • Tip is locked and sealed in place until ejected − no dropped tips that waste valuable samples
  • Two operating options for every application: Step based Matrix or Pre-sets with settings for most common applications
  • Five user profiles, personalised user interface and password protection offer unified operation for multiple users and environments
  • Ability to store up to 30 common protocols with a specific name and calibration in the Programs function
  • Service and calibration trackers remind users when the pipette requires service or repair
  • Lithium-ion battery ensures long lasting performance; unit may be recharged by a charging stand or plug
  • Pipettes can be programmed via PC by connecting to the new My Pipette™ Creator app via Bluetooth* or USB connection
Objednávacie informácie: Supplied with universal power supply. 2 year extended warranty only available via web registration.
Pozor: ClipTip 384 and ClipTip 12,5 µl tips and compatible E1-ClipTip models do not have interlocking technology.
* Bluetooth models are not available in all countries.